All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H23B
Molecular Weight:
178.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
SMILES string
B(C1CCCCC1)C2CCCCC2
InChI
1S/C12H23B/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h11-13H,1-10H2
InChI key
XNYOSXARXANYPB-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Pyr. Sol. 1 - Skin Corr. 1B
Storage Class
4.2 - Pyrophoric and self-heating hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service