Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF29667190

Silicon carbide

powder, mean particle size 0.1-1 micron, weight 50 g, purity 99%

Synonym(s):

SI676021

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23
Product GF29667190 is not currently sold in your country. Contact Technical Service

assay

99%

form

powder

manufacturer/tradename

Goodfellow 296-671-90

weight

50 g

mean particle size

0.1-1 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service