Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E27750

Sigma-Aldrich

Ethylene trithiocarbonate

97%

Synonym(s):

1,3-Dithiolane-2-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4S3
CAS Number:
Molecular Weight:
136.26
Beilstein/REAXYS Number:
106330
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

bp

307 °C (lit.)

mp

34-36 °C (lit.)

SMILES string

S=C1SCCS1

InChI

1S/C3H4S3/c4-3-5-1-2-6-3/h1-2H2

InChI key

XCWPBWWTGHQKDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

325.4 °F - closed cup

flash_point_c

163 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service