Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C005

Sigma-Aldrich

3-Quinuclidinylxanthene-9-carboxylic acid hemioxalate salt

Synonym(s):

QNX hemioxalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H21NO3 · (0.5)C2H2O4
CAS Number:
Molecular Weight:
380.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

195-198 °C

storage temp.

2-8°C

SMILES string

OC(=O)C(O)=O.O=C(OC1CN2CC[C@@H]1CC2)C3c4ccccc4Oc5ccccc35.O=C(OC6CN7CC[C@@H]6CC7)C8c9ccccc9Oc%10ccccc8%10

InChI

1S/2C21H21NO3.C2H2O4/c2*23-21(25-19-13-22-11-9-14(19)10-12-22)20-15-5-1-3-7-17(15)24-18-8-4-2-6-16(18)20;3-1(4)2(5)6/h2*1-8,14,19-20H,9-13H2;(H,3,4)(H,5,6)

InChI key

KBKSAAZOXNEJTC-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service