Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00424

Sigma-Aldrich

2-Naphthalenyl ester fluorosulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7FO3S
CAS Number:
Molecular Weight:
226.22
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

SMILES string

O=S(OC1=CC=C2C(C=CC=C2)=C1)(F)=O

InChI

1S/C10H7FO3S/c11-15(12,13)14-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H

InChI key

IJQQGAGYVGLLJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Naphthalenyl ester fluorosulfuric acid is a diaryl fluorosulfate (diaryl-OSO2F). It can be synthesized with 99% yield from the reaction between phenol and sulfuryl fluoride in the presence of triethylamine. It undergoes Suzuki-Miyaura reaction with aryl boronic acid in the presence of Pd(OAc)2 and Et3N to afford the corresponding biaryl derivative.[1]

Application

The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiaobin Liang et al.
Organic letters, 17(8), 1942-1945 (2015-04-10)
Aryl fluorosulfates were prepared by a simple method and employed as coupling partners in the Suzuki-Miyaura reaction. The cross-coupling reactions were performed in water under air at room temperature without ligands or additives such as surfactants or phase-transfer reagents and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service