Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

907413

Sigma-Aldrich

Copper inkjet ink

Synonym(s):

Cu nano ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu
UNSPSC Code:
12352200

description

Cu(0) in Cu nanoparticles: >95%

form

liquid

composition

Triethylene glycol monomethyl ether (solvent, TGME)

particle size distribution

120 nm (D90 (by Lumisizer))
50 nm (D50)

surface tension

34 dyn/cm (Pendant Drop)

viscosity

32 cP (Brookfield, Cone Spindle 40, 25ºC)

density

1.85 g/mL

cation traces

Cu: 50 wt. %

storage temp.

15-25°C

Application

This product is a conductive ink based on single-crystal copper nanoparticles in triethylene glycol monomethyl ether (TGME), is suitable for various digital printing technologies such as Inkjet and Aerosol systems. The ink offers high copper loading, low viscosity, reliable jetting with high open time, good printability, long shelf life, storage at room temperature (under Argon). Printed and laser sintered patterns provide low electrical resistivity and good adhesion to substrates.

  • Compatible printheads
KM1024/i, Ricoh E3, DMC-11610, Air Brush

  • Product fit for:
Digital Printing (Inkjet, Aerosol)
Printed Electronics:
FPD, RFID, PCB

Caution

Mix well before use.
Handle under Ar.

Physical properties

Electrical Properties
  • Sintering conditions (on glass) :
Laser sintering
Resistivity after laser sintering : ≤5 μOhm·cm (≤3 bulk)
Thermal 300 Celsius/30 min (under Argon) : ≤120 μOhm.cm (≤70 bulk)

  • Adhesion to (tested) :
Kapton®, FR4, ITO, glass
Adhesion depends on substrate, sintering conditions, substrate, pretreatment and pattern thickness (tested according to ASTM-3359-09 or ISO-2409). Substrates listed here were tested and perform well. Other substrates may also be applicable

  • Compatible printheads:
KM1024, KM1024i, Ricoh E3
Printheads listed here were tested and perform well. Other printheads may also be applicable

Storage and Stability

Store in a dry place at room temperature; Avoid exposure to direct, strong light; Do no heat above 60 °C

Legal Information

Kapton is a registered trademark of E. I. du Pont de Nemours and Company
Lumisizer is a trademark of LUM GmbH

pictograms

FlameEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Self-heat. 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 2

flash_point_f

>230.0 °F

flash_point_c

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service