Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

901359

Sigma-Aldrich

Poly(ethylene glycol) linear dendron

NHBoc terminated, PEG Mn 2000, generation 1

Synonym(s):

Dendron functionalized PEG, PFD-G1-PEG2k-NHBoc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C112H218N2O55
Molecular Weight:
2472.91
UNSPSC Code:
51171641
NACRES:
NA.23

form

solid

mol wt

PEG Mn 2000

color

white to off-white

storage temp.

−20°C

General description

  • Polymer architecture: PEG-linear dendron
  • Generation: 1
  • PEG Average Mn: 2,000
  • End group functionality: Boc-protected amine
  • No. surface groups: 2
  • Calculated mol. wt.: 2472.94 g/mol
  • Boc: tert-Butyloxycarbonyl
  • Bis-MPA: 2,2-Bis(hydroxymethyl)propionic acid

application

Bis-MPA (or 2,2-bis(methylol)propionic acid) is an aliphatic, pro-chiral molecule comprised of two hydroxyls and one carboxylic group that has been used in the synthesis of many different types of polymers. Dendrons synthesized from bis-MPA are biodegradable and have low cytotoxicity, making them well suited for use in biological research applications. These materials have been used in many applications such as signal amplification in bioassays and in drug delivery applications. PEG-core dendrons feature excellent solubility in water and a high degree of functionality per PEG chain, making them strong candidates for advanced drug delivery systems or as targeting vessels. Additionally, the terminal amine groups can readily be used in EDC or DCC coupling reactions (after Boc deprotection) with carbonyl-containing compounds, to yield highly functionalized materials for a variety of biomedical applications.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanjun Fu et al.
Bioconjugate chemistry, 17(4), 1043-1056 (2006-07-20)
The purpose of this study was to design, synthesize, and initially characterize a representative set of novel constructs for large-molecular radiographic/computed tomography (CT) contrast agents, intended for a primarily intravascular distribution. A new assembly of well-known and biocompatible components consists
Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service