Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900719

Sigma-Aldrich

Tanaka-Yu template

≥95%

Synonym(s):

Catalytic bifunctional template

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H26FN5O4Pd
CAS Number:
Molecular Weight:
670.00
UNSPSC Code:
12161600

assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Asymmetric synthesis
reaction type: C-C Bond Formation
reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

functional group

fluoro
nitrile

storage temp.

2-8°C

Application

Recyclable, catalytic, and bifunctional template developed by Jin-Quan Yu′s lab for remote site-selective C-H activation. By binding reversibly, the template is compatible with heterocycles, including quinoline derivatives, void of functional groups needed for covalent attachment.

related product

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service