Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900718

Sigma-Aldrich

N,N′-(2,3-Dimethylbutane-2,3-diyl)bis(pyridine-3-sulfonamide)

≥95%

Synonym(s):

Catalytic bifunctional template

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N4O4S2
Molecular Weight:
398.50
UNSPSC Code:
12352200

assay

≥95%

form

powder or crystals

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

functional group

amine

storage temp.

2-8°C

Application

Catalytic, bifunctional template developed by Jin-Quan Yu′s lab for remote site-selective C-H activation. By binding reversibly, the template is compatible with heterocycles void of functional groups needed for covalent attachment.

related product

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhipeng Zhang et al.
Nature, 543(7646), 538-542 (2017-03-09)
In chemical syntheses, the activation of carbon-hydrogen (C-H) bonds converts them directly into carbon-carbon or carbon-heteroatom bonds without requiring any prior functionalization. C-H activation can thus substantially reduce the number of steps involved in a synthesis. A single specific C-H

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service