900435
YYG 546 200 isiphor
Synonym(s):
Cerium activated aluminium garnet, Rare earth doped phosphor, Yellow phosphor
About This Item
Recommended Products
form
powder
composition
Halogen free, Br <700 ppm, Cl <700 ppm, Br+Cl <1000 ppm
particle size
12-14 μm (D10)
19-21 μm (D50)
26-28 μm (D90)
Application
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service