MilliporeSigma
Advances in Phosphors for Light-emitting Diodes.
The journal of physical chemistry letters (2011-06-02)
Chun Che Lin, Ru-Shi Liu
ABSTRACT
Light-emitting diodes (LEDs) are excellent candidates for general lighting because of their rapidly improving efficiency, durability, and reliability, their usability in products of various sizes, and their environmentally friendly constituents. Effective lighting devices can be realized by combining one or more phosphor materials with chips. Accordingly, it is very important that the architecture of phosphors be developed. Although numerous phosphors have been proposed in the past several years, the range of phosphors that are suitable for LEDs is limited. This work describes recent progress in our understanding of the prescription, morphology, structure, spectrum, and packaging of such phosphors. It suggests avenues for further development and the scientific challenges that must be overcome before phosphors can be practically applied in LEDs.