Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

900347

Sigma-Aldrich

Diisopropylamine borane

95%

Synonym(s):

DIPAB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H18BN
CAS Number:
Molecular Weight:
115.02
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

description

Physical form: solid, semi-solid, or liquid

assay

95%

form

solid

reaction suitability

reagent type: reductant

refractive index

n/D 1.452

density

0.790 g/mL

functional group

amine

storage temp.

2-8°C

SMILES string

CC(C)[N+]([H])([BH3-])C(C)C

InChI

1S/C6H18BN/c1-5(2)8(7)6(3)4/h5-6,8H,1-4,7H3

InChI key

CECHXEJDHZCHDL-UHFFFAOYSA-N

Application

DIPAB, together with DCAB, were developed as cost effective and easy to handle borylation reagents by the Pucheault Group at the Universite de Bordeaux. From aryl and vinyl halide electrophiles, you can easily access the boronic acid derivative of your choice, including BF3K, Bpin and MIDAs.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

162.0 °F

flash_point_c

72.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service