Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

802786

Sigma-Aldrich

Aquivion® E87-12S

membrane sheet, stabilized CF3 polymer chain ends, PFSA eq. wt. 870 g/mole SO3H, L × W × thickness 18 cm × 18 cm × 120 μm

Synonym(s):

Aquivion® SO3H, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CF2CF(OCF2CF2SO3H)]m[CF2CF2]n
CAS Number:
UNSPSC Code:
26111700
NACRES:
NA.23

form

membrane sheet

L × W × thickness

18 cm × 18 cm × 120 μm

Looking for similar products? Visit Product Comparison Guide

General description

Aquivion® PFSA ionomer membranes are melt-extruded films based on the short-side-chain (SSC) copolymer of Tetrafluoroethylene and the Sulfonyl Fluoride Vinyl Ether (SFVE) F2C=CF-O-(CF2)2-SO2F industrially produced by Solvay Specialty Polymers. Following a film hydrolysis the perfluoropolymer′s functional groups are operative in their sulfonic acid form, SO3H.

Application

Aquivion® PFSA membranes are used for electrochemical applications such as, but not limited to, polymer electrolyte fuel cells, electro-deionization systems, ozone generators, water electrolyzers, hydrogen separators and compressors, redox flow batteries as well as pervaporation or gas humidification systems.

Other Notes

Membranes are packed and sealed in a barrier protection pouch inside a cardboard box to prevent damage or deterioration. It is recommendable storing the product in a clean, humidity-controlled environment protected from direct sunlight or other sources of irradiation or heat. Sheet dimensions are based on product conditioned at 23°C and about 50% relative humidity. They may slightly vary upon exposure to different ambient conditions. Maximum temperature rating is 230°C, overheating will result in polymer degradation, discoloration and release of toxic volatile decomposition products.

Legal Information

Aquivion is a registered trademark of Syensqo Group

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service