Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

799688

Sigma-Aldrich

White-Paradine catalyst

Synonym(s):

Manganese 2,9,16,23-tetra-tert-butyl-29H,31H-phthalocyanine chloride, [2,9,16,23-Tetra-tert-butyl-29H,31H-phthalocyanine]manganese(III) chloride, [Mn(t-BuPc)]Cl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H48ClMnN8
Molecular Weight:
827.34
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

CC(C)(C)C(C=C1)=CC2=C1C3=N/C2=N\C4=C5C(C=C(C(C)(C)C)C=C5)=C6N4[Mn](Cl)N7/C(C(C=C(C(C)(C)C)C=C8)=C8/C7=N/C(C9=C/%10C=CC(C(C)(C)C)=C9)=NC%10=N/6)=N\3

InChI

1S/C48H48N8.ClH.Mn/c1-45(2,3)25-13-17-29-33(21-25)41-49-37(29)54-42-35-23-27(47(7,8)9)15-19-31(35)39(51-42)56-44-36-24-28(48(10,11)12)16-20-32(36)40(52-44)55-43-34-22-26(46(4,5)6)14-18-30(34)38(50-43)53-41;;/h13-24H,1-12H3;1H;/q-2;;+3/p-1

InChI key

NRPFPIHGBUOGKJ-UHFFFAOYSA-M

Application

The following manganese complex is a catalyst for chemoselective intramolecular C(sp3)—H amination originally reported by the White research group.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paradine, SM, et al.
Nature Chemistry (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service