Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798975

Sigma-Aldrich

Trovitch Ni-Precatalyst

Synonym(s):

(Ph2PPrDI)Ni

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H38N2NiP2
CAS Number:
Molecular Weight:
595.32
UNSPSC Code:
12161600
PubChem Substance ID:

form

powder

reaction suitability

core: nickel
reagent type: catalyst

storage temp.

2-8°C

SMILES string

CC(/C(C)=N/CCCP(C1=CC=CC=C1)C2=CC=CC=C2)=N\CCCP(C3=CC=CC=C3)C4=CC=CC=C4.[Ni]

InChI

1S/C34H38N2P2.Ni/c1-29(35-25-15-27-37(31-17-7-3-8-18-31)32-19-9-4-10-20-32)30(2)36-26-16-28-38(33-21-11-5-12-22-33)34-23-13-6-14-24-34;/h3-14,17-24H,15-16,25-28H2,1-2H3;/b35-29+,36-30+;

Inchi Key

ILRLBVPENGSFTO-CAKVIZMISA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Related Content

The Trovitch laboratory is working to develop first-row transition metal catalysts that can be used to achieve a variety of organic transformations. The first complexes from this group made available through Sigma-Aldrich are the hydrosilylation catalysts, (Ph2PPrPDI)Mn and (Ph2PPrDI)Ni. Although these compounds are formally zerovalent, both feature a reduced chelate that coordinates to the metal center beyond its redox non-innocent core.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service