Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798487

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide)-COOH

Mw 17,000, lactide:glycolide 85:15

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)x(C2H2O2)y-1C3H6O2
UNSPSC Code:
12162002

form

powder

feed ratio

lactide:glycolide 85:15

mol wt

Mw 17,000

storage temp.

2-8°C

General description

Polymers with free -COOH groups are more hydrophilic (e.g., R503H compared to R503)

Application

This copolymer can be used in targeted drug delivery. The carboxylic acid can be used for coordination or ligation or further reactions.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service