Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775991

Sigma-Aldrich

Poly(L-lactide), acrylate terminated

average Mn 2,500, PDI ≤1.2

Synonym(s):

PLA, PLLA acrylate, acrylate-PLA-OH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)nC5H8O3
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 2,500

PDI

≤1.2

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

This poly(L-lactide) is a well-defined biodegradable polymer which is functionalized with an alcohol on one end and an acrylate on the other. Acrylates are commonly used in polymerizations or in ligatation to thiols with thiol-ene click chemistry. Polylactide is commonly used in several applications including drug delivery, compatibilizer, emulsifier, thermoplastic elastomer or impact-resistant plastic.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadege Handke;
Journal of Polymer Science Part A: Polymer Chemistry, 49, 1341-1350 (2011)
Synthesis of Polymer Network Scaffolds from L-Lactide and Poly(ethylene glycol) and Their Interaction with Cells
Dong Keun Han;
Macromolecules, 30(20) (1997)

Articles

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service