Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

774553

Sigma-Aldrich

Pentafluorophenyl 4-vinylbenzoate

99% (HPLC)

Synonym(s):

4-styrene pentafluorophenoate, PFP ester monomer, activated ester styrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H7F5O2
CAS Number:
Molecular Weight:
314.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

99% (HPLC)

form

solid

mp

39-44 °C

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(OC(=O)c2ccc(C=C)cc2)c(F)c1F

InChI

1S/C15H7F5O2/c1-2-7-3-5-8(6-4-7)15(21)22-14-12(19)10(17)9(16)11(18)13(14)20/h2-6H,1H2

InChI key

QLSHRSPPXSAGTR-UHFFFAOYSA-N

Application

This styrene monomer contains a highly reactive "activated ester" for coupling reactions with alcohols or amines prior to polymerization; allowing for easy use in making customized architectures.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nilles; et al.
Journal of Polymer Science Part A: Polymer Chemistry, 47, 1696-1705 (2009)
Jinhwa Seo et al.
Langmuir : the ACS journal of surfaces and colloids, 26(3), 1830-1836 (2009-09-19)
We demonstrate that chemically stable, multifunctional polymer thin films can be obtained using the layer-by-layer (LbL) deposition based on covalent bonds between adsorbing chains. Poly(pentafluorophenyl-4-vinylbenzoate) (P1) or poly(pentafluorophenylacrylate) (P2) polymers were assembled with poly(allyl amine) (PAAm) to yield LbL multilayer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service