Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771643

Sigma-Aldrich

Gold nanorods

25 nm diameter, λmax, 550 nm, dispersion in H2O

Synonym(s):

Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

colloidal suspension
dispersion in H2O
nanorod

contains

CTAB as stabilizer

concentration

>45 μg/mL in H2O

diam. × L

25 nm × 34 nm (±10%)

diameter

25 nm

λmax

550 nm

storage temp.

2-8°C

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanoparticles have unique properties like biocompatibility, non-cytotoxicity, good optical and electronic properties. They are widely in targeted drug delivery, bioassays and polymer-based sensors. [1]Gold nanorods exhibits a transverse plasmon resonance in the wavelength range of around 520 nm and longitudinal plasmon resonance in the wavelength range of greater than 600 nm which makes it very attractive for optical imaging in tissue since light in this wavelength range can penetrate deep into the tissue due to low absorption and scattering.[2]

Application

Used in cancer nano-therapy.[3]

Features and Benefits

Strong light absorption, localized surface plasmon resonance, quantum effect, biocompatibility, low toxicity, consistency, easy synthesis, ability to be readily functionalized with targeting ligands.[3]

Legal Information

This product was produced under the methods claimed in U.S. Pat. Nos. 8,956,440 and 8,241,922

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raja Gopal Rayavarapu et al.
International journal of biomedical imaging, 2007, 29817-29817 (2008-03-21)
We have synthesized and characterized gold nanoparticles (spheres and rods) with optical extinction bands within the "optical imaging window." The intense plasmon resonant driven absorption and scattering peaks of these nanoparticles make them suitable as contrast agents for optical imaging
Nanoscopic Study of Gold Nanorods Used for Efficient Delivery of Sirna in Cancer Nano-Therapy
Tahmasebifar, A., Yazdani, Y., Gill, P., & Shahbazi, M.
American Journal of Advanced Drug Delivery, 2(2), 254-269 (2014)
Scott H Brewer et al.
Langmuir : the ACS journal of surfaces and colloids, 21(20), 9303-9307 (2005-09-21)
The interaction of bovine serum albumin (BSA) with gold colloids and surfaces was studied using zeta-potential and quartz crystal microbalance (QCM) measurements, respectively, to determine the surface charge and coverage. The combination of these two measurements suggests that BSA binding

Articles

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service