Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767956

Sigma-Aldrich

4-Fluorobenzylphosphonic acid

99% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8FO3P
CAS Number:
Molecular Weight:
190.11
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99% (HPLC)

form

solid

mp

176-180 °C

SMILES string

FC1=CC=C(CP(O)(O)=O)C=C1

InChI

1S/C7H8FO3P/c8-7-3-1-6(2-4-7)5-12(9,10)11/h1-4H,5H2,(H2,9,10,11)

InChI key

RHUKKXTWNWQHJT-UHFFFAOYSA-N

Application

Self-assembly material is used to chemically modify transparent conducting oxides; tune the effective surface work function; and control the surface energy and wettability for OLEDs and OPVs application.[1][2][3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tailoring the work function of indium tin oxide electrodes in electrophosphorescent organic light-emitting diodes.
Sharma A, et al.
Journal of Applied Physics, 105(8), 084507-084507 (2009)
Effect of phosphonic acid surface modifiers on the work function of indium tin oxide and on the charge injection barrier into organic single-layer diodes.
Sharma A, et al.
Journal of Applied Physics, 105(7), 074511-074511 (2009)
Tuning the Work Function of Polar Zinc Oxide Surfaces using Modified Phosphonic Acid Self-Assembled Monolayers.
Lange I, et al.
Advances in Functional Materials, 24(44), 7014-7024 (2014)
Nicholas J Morris et al.
Nanoscale, 11(37), 17308-17318 (2019-09-13)
Eutectic gallium-indium (EGaIn) is a room temperature liquid metal that can be readily fabricated into nanoparticles which naturally form a thin, passivating gallium oxide shell. These core-shell nanoparticles are of interest for a variety of stimuli-responsive applications, which often utilize

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service