Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

757209

Sigma-Aldrich

3-Phenylisoquinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N
CAS Number:
Molecular Weight:
205.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

powder or crystals

mp

100-105 °C

SMILES string

c1ccc(cc1)-c2cc3ccccc3cn2

InChI

1S/C15H11N/c1-2-6-12(7-3-1)15-10-13-8-4-5-9-14(13)11-16-15/h1-11H

InChI key

RBJOTRVJJWIIER-UHFFFAOYSA-N

General description

3-phenylisoquinoline can be prepared by the Bradsher procedure.[1] It is an important component in the essential oils obtained from Ageratum conyzoides. 2

Application

This material can be used as a ligand to synthesize a variety of OLED dopants.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An unexpected product during synthesis of 3-phenylisoquinoline: Improved preparation of 4-hydroxy-3-phenylisoquinoline
Sard H
Journal of Heterocyclic Chemistry null
An unexpected product during synthesis of 3-phenylisoquinoline: Improved preparation of 4-hydroxy-3-phenylisoquinoline
Sard H
Journal of Heterocyclic Chemistry, 1085-1087 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service