Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751898

Sigma-Aldrich

Lithium chloride

anhydrous, chunks, 99.99% trace metals basis

Synonym(s):

Lithium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
99.99% trace metals basis
grade:
anhydrous
form:
chunks
solubility:
H2O: soluble

grade

anhydrous

assay

99.99% trace metals basis

form

chunks

mp

605 °C (lit.)

solubility

H2O: soluble

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium chloride is used in chemical synthesis[1] as well as in the separation of RNA in biological system[2]. Lithium chloride can also be used as a starting material for the formation of battery material[3].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Efficient synthesis of functionalized organozinc compounds by the direct insertion of zinc into organic iodides and bromides.
Arkady Krasovskiy et al.
Angewandte Chemie (International ed. in English), 45(36), 6040-6044 (2006-08-11)
RNA isolation from siliques, dry seeds, and other tissues of Arabidopsis thaliana.
Yuji Suzuki et al.
BioTechniques, 37(4), 542-542 (2004-11-03)
Wu, X. L.; et al.
Advanced Materials, 21, 2710-2714 (2009)
Alessandro Di Maio et al.
EvoDevo, 6, 17-17 (2015-07-15)
Wnt signaling is one of the earliest and most highly conserved regulatory pathways for the establishment of the body axes during regeneration and early development. In regeneration, body axes determination occurs independently of tissue rearrangement and early developmental cues. Modulation
A M Petrosyan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 105, 623-625 (2013-02-07)
We argue that the recently reported crystals "L-threonine formate" as well as "L-alanine lithium chloride" and "bis L-alanine lithium chloride" actually are the well-known crystals L-threonine and L-alanine, respectively.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service