Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749699

Sigma-Aldrich

Propylcarboxylic acid functionalized silica

mesoporous, nanoparticles, TCP-porphine labeled, 200 nm particle size, pore size 4 nm

Synonym(s):

Silica nanoparticles, mesoporous, propylcarboxylic acid functionalized, meso-Tetra(4-carboxyphenyl)porphine labeled

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

form

mesoporous
nanoparticles
powder (approximately spherical)

particle size

200 nm

pore size

4 nm pore size

bp

2230 °C (lit.)

mp

1600-1725 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

These well defined mesoporous materials are used in applications that include separations,[1] drug delivery,[2] optical imaging,[3] and composites.[4]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang, D.; Wang, X.; Qiao, Z.; Tang, D.; Liu, Y.; Huo, Q.
The Journal of Physical Chemistry C, 114, 12505-12505 (2010)
Thiam-Leng Chew et al.
Advances in colloid and interface science, 153(1-2), 43-57 (2010-01-12)
Separation of carbon dioxide (CO(2)) from gaseous mixture is an important issue for the removal of CO(2) in natural gas processing and power plants. The ordered mesoporous silicas (OMS) with uniform pore structure and high density of silanol groups, have
Huan Meng et al.
ACS nano, 4(8), 4539-4550 (2010-08-25)
Overexpression of drug efflux transporters such as P-glycoprotein (Pgp) protein is one of the major mechanisms for multiple drug resistance (MDR) in cancer cells. A new approach to overcome MDR is to use a co-delivery strategy that utilizes a siRNA
Wang, Z.; Stein, A.
Chemistry of Materials, 20, 1029-1029 (2008)

Articles

Mesoporous Materials include a range of high surface area porous silicates with applications in gas adsorption, drug delivery, diagnostics and catalysis.

Mesoporous silica used in wastewater remediation, air cleaning, catalysis, bio-catalysis, and drug delivery.

Silica's versatility spans various industries, including biomedical applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service