Key Documents
744077
Sigma-Aldrich
COware
screwable two-chamber glass sytem
About This Item
UNSPSC Code:
12352000
General description
Two-chamber glass equipment ideal for a reaction scale of 0.1-1 mmol (total volume 20 mL), two Screw caps, two stabilizing PTFE discs (recyclable). Ten single use silicone/PTFE septa (in combination with the stabilizing discs each septum allows for a maximum of 2 piercings using 21G/Ø0.80 mm syringes or smaller).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Articles
What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.