Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

741647

Sigma-Aldrich

Chloromethyl 2-ethylbutyrate

95%

Synonym(s):

2-Ethylbutyric acid chloromethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13ClO2
CAS Number:
Molecular Weight:
164.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.427

density

1.031 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCC(CC)C(=O)OCCl

InChI

1S/C7H13ClO2/c1-3-6(4-2)7(9)10-5-8/h6H,3-5H2,1-2H3

InChI key

AAACXXOAKLEEOX-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

145.9 °F

flash_point_c

63.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service