Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735752

Sigma-Aldrich

(S)-1-Boc-3-(aminomethyl)piperidine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O2
Molecular Weight:
214.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.4754

density

1.008 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCC[C@@H](CN)C1

InChI

1S/C11H22N2O2/c1-11(2,3)15-10(14)13-6-4-5-9(7-12)8-13/h9H,4-8,12H2,1-3H3/t9-/m0/s1

InChI key

WPWXYQIMXTUMJB-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service