Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735418

Sigma-Aldrich

Ethyl (R)-pyrrolidine-3-carboxylate hydrochloride

95%

Synonym(s):

(R)-Pyrrolidine-3-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14ClNO2
Molecular Weight:
179.64
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

optical activity

[α]/D -15.0°, c = 0.5% in H2O

mp

230-270 °C

storage temp.

2-8°C

SMILES string

Cl.CCOC(=O)[C@@H]1CCNC1

InChI

1S/C7H13NO2.ClH/c1-2-10-7(9)6-3-4-8-5-6;/h6,8H,2-5H2,1H3;1H/t6-;/m1./s1

InChI key

AYYKCALHTPKNOI-FYZOBXCZSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service