Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

734241

Sigma-Aldrich

1-Iodo-2,4-dimethyl-6-nitrobenzene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$30.70

$30.70


Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$30.70

About This Item

Empirical Formula (Hill Notation):
C8H8INO2
CAS Number:
Molecular Weight:
277.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$30.70


Available to ship onApril 29, 2025Details


Request a Bulk Order

Quality Level

assay

97%

form

solid

mp

102-107 °C

functional group

iodo
nitro

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(I)c(c1)[N+]([O-])=O

InChI

1S/C8H8INO2/c1-5-3-6(2)8(9)7(4-5)10(11)12/h3-4H,1-2H3

InChI key

IHFAHQKEKVTXOG-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service