Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

729272

Sigma-Aldrich

Zinc chloride tetrahydrofuran complex (1:2)

97%

Synonym(s):

Dichlorobis(tetrahydrofuran)zinc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Zn · 2C4H8O
CAS Number:
Molecular Weight:
280.51
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

core: zinc
reagent type: catalyst

mp

113-118 °C

SMILES string

Cl[Zn]Cl.C1CCOC1.C2CCOC2

InChI

1S/2C4H8O.2ClH.Zn/c2*1-2-4-5-3-1;;;/h2*1-4H2;2*1H;/q;;;;+2/p-2

InChI key

YKZBFJVZMFIZLW-UHFFFAOYSA-L

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

nwg

flash_point_f

171.0 °F

flash_point_c

77.22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service