Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727776

Sigma-Aldrich

Trityl-OH ChemMatrix® resin

35-100 mesh particle size (wet sieved)

Synonym(s):

4-(Diphenylhydroxymethyl)benzoic acid, polymer-bound to ChemMatrix® resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product line

ChemMatrix®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.30-0.60 mmol/g loading

particle size

35-100 mesh (wet sieved)

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-8°C

Application

Trityl-OH ChemMatrix® resin can be used as efficient support in solid-phase peptide synthesis (SPPS). It is also be used to prepare novel hydroxylamine ChemMatrix resin, which is used for the synthesis of polypeptides containing aminohydroxamic acids.[1][2]

Legal Information

ChemMatrix is a registered trademark of Matrix Innovation, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid phase synthesis of peptide hydroxamic acids on poly (ethylene glycol)-based support
Cal Marta, et al.
Journal of Peptide Science, 19(1), 9-15 (2013)
A continuous flow protocol to generate, regenerate, load, and recycle chlorotrityl functionalised resins
Spare LK, et al.
Reaction Chemistry & Engineering, 4(7), 1309-1317 (2019)

Articles

ChemMatrix® is a proprietary, 100% PEG (polyethylene glycol) based resin from PCAS BioMatrix.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service