Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727652

Sigma-Aldrich

(RS)-3-Pyrrolidinemethanol

≥98% (GC)

Synonym(s):

(RS)-3-(Hydroxymethyl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein/REAXYS Number:
1420904
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (GC)

form

liquid

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OCC1CCNC1

InChI

1S/C5H11NO/c7-4-5-1-2-6-3-5/h5-7H,1-4H2

InChI key

QOTUIIJRVXKSJU-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

186.8 °F - closed cup

flash_point_c

86 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service