Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720607

Sigma-Aldrich

(S)-(−)-2-Butyl-CBS-oxazaborolidine solution

1.0 M in toluene

Synonym(s):

(S)-1-Butyl-3,3-diphenylhexahydropyrrolo[1,2-c][1,3,2]oxazaborole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26BNO
Molecular Weight:
319.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

liquid

optical activity

[α]22/D −72.0°, c = neat neat

concentration

1.0 M in toluene

refractive index

n20/D 1.514

density

0.916 g/mL at 25 °C

functional group

phenyl

SMILES string

CCCCB1OC([C@@H]2CCCN12)(c3ccccc3)c4ccccc4

InChI

1S/C21H26BNO/c1-2-3-16-22-23-17-10-15-20(23)21(24-22,18-11-6-4-7-12-18)19-13-8-5-9-14-19/h4-9,11-14,20H,2-3,10,15-17H2,1H3/t20-/m0/s1

InChI key

FUVQOWKGWXFWHP-FQEVSTJZSA-N

General description

(S)-2-Butyl-CBS-oxazaborolidine is a Corey−Bakshi−Shibata (CBS) reagent commonly used in the enantioselective reduction of ketones.[1][2][3]

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

50.0 °F - open cup

flash_point_c

10 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Strategy towards the enantioselective synthesis of schiglautone A
Le Chapelain C
Organic & Biomolecular Chemistry, 15(29), 6242-6256 (2017)
Total Synthesis of Four Stereoisomers of (4 Z, 7 Z, 10 Z, 12 E, 16 Z, 18 E)-14, 20-Dihydroxy-4, 7, 10, 12, 16, 18-docosahexaenoic Acid and Their Anti-inflammatory Activities
Goto T, et al.
The Journal of Organic Chemistry, 80(15), 7713-7726 (2015)
New synthetic route to the C. 14- C. 38 segment of halichondrins
Stamos DP, et al.
The Journal of Organic Chemistry, 62(22), 7552-7553 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service