Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

711349

Sigma-Aldrich

Organic photovoltaic ink system

Synonym(s):

OPV ink system, Plexcore® PV 1000, Plexcore® PV ink system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103

form

liquid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Plextronics, Inc. U.S. Patent 6,166,172.
Plexcore is a registered trademark of Plextronics, Inc.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service