Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

709107

Sigma-Aldrich

Copper in charcoal

3 wt. %

Synonym(s):

Nanoparticulate Cu/C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu/C
UNSPSC Code:
12352300
NACRES:
NA.22

form

powder

reaction suitability

core: copper
reagent type: catalyst

concentration

3 wt. %

Application

A heterogeneous catalyst, primarily reported to mediate a tandem diazo transfer-click reaction starting from the corresponding amine rather than the azide.  This catalyst system has been displayed to promote this one-pot reaction series for several examples and is also easily recyclable.
Catalyst for one-pot diazo transfer-click reactions

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Tien Lee,
Advanced Synthesis & Catalysis, 351(18), 3139-3142 (2009)

Related Content

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service