Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

703532

Sigma-Aldrich

Magnesium bis(diisopropyl)amide solution

0.70 M in THF

Synonym(s):

Magnesium diisopropylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg[((CH3)2CH)2N]2
CAS Number:
Molecular Weight:
224.67
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

form

liquid

Quality Level

concentration

0.70 M in THF
18 % (w/w)

density

0.871 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)N([Mg]N(C(C)C)C(C)C)C(C)C

InChI

1S/2C6H14N.Mg/c2*1-5(2)7-6(3)4;/h2*5-6H,1-4H3;/q2*-1;+2

InChI key

XDBOBNVQEBSKFO-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1A - STOT SE 3 - Water-react 2

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

<-13.0 °F

flash_point_c

< -25 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service