Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

695556

Sigma-Aldrich

3-Phenylaziridine-2-carboxaldehyde dimer

Synonym(s):

6-Phenyl-2-(3-phenyl-2-aziridinyl)-3-oxa-1-azabicyclo[3.1.0]hexan-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O2
CAS Number:
Molecular Weight:
294.35
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

132-136 °C

functional group

aldehyde
phenyl

SMILES string

O=C[C@H]1N[C@@H]1c2ccccc2

InChI

1S/C9H9NO/c11-6-8-9(10-8)7-4-2-1-3-5-7/h1-6,8-10H/t8-,9-/m1/s1

InChI key

ROCQNVNJGDESAQ-RKDXNWHRSA-N

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Yudin laboratory is known for the development of amphoteric molecules and their application in synthesis. The corresponding reagents possess nucleophilic and electrophilic functional groups that do not prematurely react with each other.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service