Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

692042

Sigma-Aldrich

3-(Trifluoromethyl)phenethyl bromide

95%

Synonym(s):

1-(2-Bromoethyl)-3-(trifluoromethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8BrF3
CAS Number:
Molecular Weight:
253.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.486

bp

92-94 °C/12 mmHg

density

1.495 g/mL at 25 °C

functional group

bromo
fluoro

SMILES string

FC(F)(F)c1cccc(CCBr)c1

InChI

1S/C9H8BrF3/c10-5-4-7-2-1-3-8(6-7)9(11,12)13/h1-3,6H,4-5H2

InChI key

AZLYAPKPZBXCGX-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service