Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

687863

Sigma-Aldrich

(11-Mercaptoundecyl)tetra(ethylene glycol) functionalized gold nanoparticles

2 % (w/v) in H2O

Synonym(s):

(1-Mercaptoundec-11-yl)tetra(ethylene glycol) functionalized gold nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

nanoparticles
solution

concentration

2 % (w/v) in H2O

particle size

3.5-5.5 nm (TEM)

solubility

H2O: soluble
alcohol: soluble

density

1.00 g/mL

functional group

hydroxyl

storage temp.

−20°C

General description

Monodispersed, water-soluble functionalized gold nanoparticles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service