Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68755

Sigma-Aldrich

Chloro-methyl-phenyl-vinylsilane

technical, ≥95% (GC)

Synonym(s):

Methyl-phenyl-vinylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11ClSi
CAS Number:
Molecular Weight:
182.72
Beilstein/REAXYS Number:
2935625
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

assay

≥95% (GC)

form

liquid

refractive index

n20/D 1.520

bp

79 °C/3 mmHg (lit.)

density

1.040 g/mL at 20 °C (lit.)

SMILES string

C[Si](Cl)(C=C)c1ccccc1

InChI

1S/C9H11ClSi/c1-3-11(2,10)9-7-5-4-6-8-9/h3-8H,1H2,2H3

InChI key

GSXJAPJSIVGONK-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service