Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68392

Sigma-Aldrich

Ammonium carbonate

tested according to NF

Synonym(s):

Ammonii carbonas, Hartshorn salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2CO3
CAS Number:
Molecular Weight:
96.09
Beilstein/REAXYS Number:
4253487
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

agency

USP
tested according to NF

vapor density

2.7 (vs air)

form

powder

SMILES string

N.N.OC(O)=O

InChI

1S/CH2O3.2H3N/c2-1(3)4;;/h(H2,2,3,4);2*1H3

InChI key

PRKQVKDSMLBJBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for N-oxidation of amines[1]

Reactant for the the synthesis of nitriles[2]

Reagent for synthesis of propylene carbonate[3]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bharathi Balagam et al.
Inorganic chemistry, 47(3), 1173-1178 (2008-01-09)
The reactivity of the peroxymonocarbonate ion, HCO4- (an active oxidant derived from the equilibrium reaction of hydrogen peroxide and bicarbonate), has been investigated in the oxidation of aliphatic amines. Tertiary aliphatic amines are oxidized to the corresponding N-oxides in high
Vikas N. Telvekar and Rajesh A. Rane
Tetrahedron Letters, 48, 6051-6053 (2007)
Shiyong Huang, et al
Catalysis Communications, 9, 276-280 (2007)
Tomohisa Hasunuma et al.
Biotechnology for biofuels, 7(1), 493-493 (2015-02-05)
To ensure reliable sources of energy and raw materials, the utilization of sustainable biomass has considerable advantages over petroleum-based energy sources. Photosynthetic algae have attracted attention as a third-generation feedstock for biofuel production, because algae cultivation does not directly compete
Mirosława Różycka et al.
PloS one, 9(12), e114308-e114308 (2014-12-10)
Fish otoliths, biominerals composed of calcium carbonate with a small amount of organic matrix, are involved in the functioning of the inner ear. Starmaker (Stm) from zebrafish (Danio rerio) was the first protein found to be capable of controlling the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service