Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

683558

Sigma-Aldrich

(S)-Mac-H

Synonym(s):

MacMillan catalyst - Hantzsch Ester combination

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C13H19NO4)6 (C8H16N2O · CF3CO2H)1
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

SMILES string

OC(=O)C(F)(F)F.CN1[C@H](NCC1=O)C(C)(C)C.CCOC(=O)C2=C(C)NC(C)=C(C2)C(=O)OCC.CCOC(=O)C3=C(C)NC(C)=C(C3)C(=O)OCC.CCOC(=O)C4=C(C)NC(C)=C(C4)C(=O)OCC.CCOC(=O)C5=C(C)NC(C)=C(C5)C(=O)OCC.CCOC(=O)C6=C(C)NC(C)=C(C6)C(=O)OCC.CCOC(=O)C7=C(C)NC(C)=C(C7)C(=O)OCC

InChI

1S/6C13H19NO4.C8H16N2O.C2HF3O2/c6*1-5-17-12(15)10-7-11(13(16)18-6-2)9(4)14-8(10)3;1-8(2,3)7-9-5-6(11)10(7)4;3-2(4,5)1(6)7/h6*14H,5-7H2,1-4H3;7,9H,5H2,1-4H3;(H,6,7)/t;;;;;;7-;/m......0./s1

InChI key

NWCRTIKSGFXFGW-KDCVMDMUSA-N

Application

A mixture of a chiral imidazolidinone with the Hantzsch ethyl ester; formally transfers hydrogen asymmetrically to enals resulting in chiral aldehydes.[1]

Legal Information

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphane G Ouellet et al.
Journal of the American Chemical Society, 127(1), 32-33 (2005-01-06)
The first enantioselective organocatalytic hydride reduction has been accomplished. The use of iminium catalysis has provided a new organocatalytic strategy for the enantioselective reduction of beta,beta-substituted alpha,beta-unsaturated aldehydes to generate beta-stereogenic aldehydes. The use of imidazolidinone 2 as the asymmetric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service