MilliporeSigma
Enantioselective organocatalytic hydride reduction.
Journal of the American Chemical Society (2005-01-06)
Stéphane G Ouellet, Jamison B Tuttle, David W C MacMillan
ABSTRACT
The first enantioselective organocatalytic hydride reduction has been accomplished. The use of iminium catalysis has provided a new organocatalytic strategy for the enantioselective reduction of beta,beta-substituted alpha,beta-unsaturated aldehydes to generate beta-stereogenic aldehydes. The use of imidazolidinone 2 as the asymmetric catalyst has been found to mediate the transfer of hydrogen to a large class of enal substrates from ethyl Hantzsch ester. The capacity of catalyst 2 to accelerate E-Z isomerization prior to selective E-olefin reduction allows the implementation of geometrically impure enals in this operationally simple protocol.
Sigma-Aldrich
(2S,5S)-(−)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone, 97%