Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681547

Sigma-Aldrich

QuadraPure® Metal Scavengers Kit II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Kit Components Only

Product No.
Description

  • QuadraPure® IMDAZ, 100-400 μm particle size, extent of labeling: 1.5 mmol/g loading, 1 % cross-linked with divinylbenzene 5 g

Kit Components Also Available Separately

Product No.
Description
SDS

  • 655422QuadraPure® TU, macroporous, 400-600 μm particle size 5 gSDS

  • 657026QuadraPure® IDA, macroporous, 350-750 μm particle size 5 gSDS

  • QuadraPure® AMPA, macroporous, 350-750 μm particle size 5 g

  • 657646QuadraPure® AEA, 100-400 μm particle size, extent of labeling: 1.3 mmol/g loading, 1 % cross-linked with divinylbenzene 5 gSDS

  • 657662QuadraPure® MPA, 100-400 μm particle size, extent of labeling: 1.5 mmol/g loading, 1 % cross-linked with divinylbenzene 5 gSDS

  • 668575QuadraPure® DET, 450-650 μm, extent of labeling: 1.0-2.0 mmol/g loading, macroporous 5 gSDS

  • 668583QuadraPure® EDA, 500-800 μm, extent of labeling: 1.5-2.0 mmol/g loading, macroporous 5 gSDS

  • 668591QuadraPure® BZA, 400-1100 μm, extent of labeling: 20 mg/g loading, macroporous 5 gSDS

  • QuadraPure® BDZ, 400-750 μm, extent of labeling: 1.0-1.4 mmol/g loading, macroporous 5 g

See All (9)

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QuadraPure Cartridges for Removal of Trace Metal from Reaction Mixtures in Flow
A. Hinchcliffe, et al.
Organic Process Research & Development, 11, 477-481 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service