Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679488

Sigma-Aldrich

Rhodium nanoparticles entrapped in aluminum hydroxide matrix preparation

5 wt. % loading

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rh/AlO(OH)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: rhodium
reagent type: catalyst

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Application

Supported Rh nanoparticle catalyst.[1]

Used for promoting photocatalytic hydrogen production[2]

Catalyst for:
  • CO oxidation[3]
  • Electrocatalytic applications[4]
  • Arene hydrogenation[5]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuya Shimura et al.
Chemical communications (Cambridge, England), 47(31), 8958-8960 (2011-07-08)
Photodeposition of a Rh cocatalyst under atmospheric conditions could simultaneously provide both Rh metal and oxide nanoparticles on a K(2)Ti(6)O(13) photocatalyst, both of which cooperatively promoted the photocatalytic hydrogen production from water and methane.
Ryan R Dykeman et al.
Inorganic chemistry, 50(3), 717-719 (2011-01-07)
The imidazolium functionalized bipyridine compounds, {4,4'-bis[7-(2,3-dimethylimidazolium)heptyl]-2,2'-bipyridine}(2+) ([BIHB](2+)) and {4,4'-bis[(1,2-dimethylimidazolium)methyl]-2,2'-bipyridine}(2+) ([BIMB](2+)), were prepared and used as Rh nanoparticle stabilizers. The dispersed Rh nanoparticles were used as catalysts in the biphasic hydrogenation of various arene substrates. The catalytic activity was strongly influenced
In Soo Park et al.
Chemical communications (Cambridge, England), (45), 5667-5669 (2005-11-18)
A new recyclable rhodium catalyst was synthesized by a simple procedure from readily available reagents, which showed high activities in the hydrogenation of various arenes under 1 atm H2 at room temperature.
V S Patil et al.
Journal of colloid and interface science, 358(1), 238-244 (2011-04-02)
Nearly monodisperse rhodium nanoparticles with adsorbed NH(x) were synthesized at the CCl(4)-water interface. The presence of NH(x)-adsorbed species was confirmed by energy-dispersive X-ray analysis (EDX) and X-ray photoelectron spectroscopy (XPS) studies. The synthesis of controlled size 2-38 nm rhodium particles
Supported rhodium oxide nanoparticles as highly active CO oxidation catalysts.
D A J Michel Ligthart et al.
Angewandte Chemie (International ed. in English), 50(23), 5306-5310 (2011-04-22)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service