Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

669393

Sigma-Aldrich

N-Methyl-O-benzoylhydroxylamine hydrochloride

97%

Synonym(s):

Benzoic acid N-hydroxymethylamine ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)CO2(NHCH3) HCl
CAS Number:
Molecular Weight:
187.62
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

133 °C (dec.)

functional group

amine
phenyl

storage temp.

2-8°C

SMILES string

[Cl-].C[NH2+]OC(=O)c1ccccc1

InChI

1S/C8H9NO2.ClH/c1-9-11-8(10)7-5-3-2-4-6-7;/h2-6,9H,1H3;1H

InChI key

FTCDQNQIGUCFSA-UHFFFAOYSA-N

application

Benzoylation Reagents Simple Tools for a-Oxygenation of Aldehydes and Ketones

Reactant for preparation of:
  • NHC-catalyzed redox esterification and cycloaddition reactions
  • Pd/pyrrolidine-catalyzed Tsuji-Trost cyclization of aldehydes
  • α-acyloxy ketones via oxyacylation reaction
Reagent for α-acyloxylation of aldehydes and ketones. The reagent is not sensitive to air and moisture.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cory S Beshara et al.
Organic letters, 7(25), 5729-5732 (2005-12-03)
[chemical reaction: see text]. A simple, one-pot method for the alpha-acyloxylation of carbonyl compounds that proceeds at room temperature in the presence of both moisture and air has been developed. Treatment of a variety of aldehydes and both cyclic and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service