Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

668923

Sigma-Aldrich

Di-μ-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino-κN)methyl]phenyl-κC]palladium dimer

97%

Synonym(s):

Nájera Catalyst I, Di-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl-C]di-palladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16Cl6N2O2Pd2
CAS Number:
Molecular Weight:
813.98
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

O\N=C(/c1ccc(Cl)cc1)c2ccc(Cl)cc2[Pd]Cl.O\N=C(/c3ccc(Cl)cc3)c4ccc(Cl)cc4[Pd]Cl

InChI

1S/2C13H8Cl2NO.2ClH.2Pd/c2*14-11-5-1-9(2-6-11)13(16-17)10-3-7-12(15)8-4-10;;;;/h2*1-3,5-8,17H;2*1H;;/q;;;;2*+1/p-2/b2*16-13+;;;;

InChI key

GIMFEGPTUHULMU-VAABLSLRSA-L

Application

Catalyst for:
  • Suzuki coupling
  • Suzuki-Miyaura coupling
  • Najera palladacycle-catalyzed intermolecular Heck reaction of Morita-Baylis-Hillman adducts
  • Hiyama cross-coupling
  • Palladium-catalyzed acylation of terminal alkynes with acid chlorides
  • Copper and amine free Sonogashira coupling reaction
Highly effective catalyst for multiple aryl vinylation via Heck coupling.[1] Palladacycle catalyst employed in a variety of carbon-carbon bond forming reactions.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

European Journal of Organic Chemistry, 3482-3482 (2005)
Synthesis, 1713-1713 (2004)

Articles

Palladacyclic catalysts by Bedford's group facilitate coupling reactions with low catalyst loadings and challenging aryl chlorides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service