663093
(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone
97%
Synonym(s):
(2R,5R)-2-tert-Butyl-3-methyl-5-phenylmethyl-4-imidazolidinone
About This Item
Recommended Products
assay
97%
form
solid
mp
93-100 °C
SMILES string
CN1[C@@H](N[C@H](Cc2ccccc2)C1=O)C(C)(C)C
InChI
1S/C15H22N2O/c1-15(2,3)14-16-12(13(18)17(14)4)10-11-8-6-5-7-9-11/h5-9,12,14,16H,10H2,1-4H3/t12-,14-/m1/s1
InChI key
SKHPYKHVYFTIOI-TZMCWYRMSA-N
General description
Application
Features and Benefits
- Superior enantiocontrol in numerous transformations
- High activities at low catalyst loadings
- Extraordinary functional group tolerance
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service