Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

660256

Sigma-Aldrich

Nitroso-15N-benzene

99 atom % 15N, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H515NO
CAS Number:
Molecular Weight:
108.10
UNSPSC Code:
12352005

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 15N

assay

97% (CP)

form

solid

SMILES string

O=[15N]C1=CC=CC=C1

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service