Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

660175

Sigma-Aldrich

Sodium silica gel Stage II

extent of labeling: ~35 wt. % Na-K loading

Synonym(s):

Na-SG-II

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
11101527
NACRES:
NA.22

form

solid

autoignition temp.

248-257 °F

reaction suitability

reagent type: reductant

extent of labeling

~35 wt. % Na-K loading

density

0.7 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Mild reducing agent capable of reducing alcohols, useful for drying and purifying organic solvents, producing hydrogen from water for hydrogenation chemistry.

Alternative to other reagents for desulfurizations, dehalogenations(as Wurtz coupling reactions), and Birch reductions.[1]

Additionally can be used as:
  • A reducing agent for aliphatic ester substrates[2]
A deprotectant in place of the typical heavy metal deprotectant[3]

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 2

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Nandi, et al.,
Tetrahedron Letters, 50, 3864-3866 (2009)
P. F. Vogt and B. Bodnar,
Spec. Chem., 29, 22-24 (2009)
Dye, J. L. et al
Journal of the American Chemical Society, 127, 9339-9339 (2005)

Articles

SiGNa Chemistry has recently developed a technology for encapsulating alkali metals into nano-structured porous oxides to create stable, free flowing powders with varying reactivities (based on the intended application).

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the difference between the NaK Silica gels (Na2K) , for example Stage I or Stage II?

    1 answer
    1. These alkali silica gels, which are offered by Sigma-Aldrich for SiGNa Chemistry, can reduce some of the handling precautions traditionally associated with alkali metals, while still retaining the reducing power of the parent alkali.They are classified into three categories. Stage 0 powders, like 660140 or 660159, are air-sensitive, but can easily be used in continuous-flow applications. Stage I powders, like 660167, are non-pyrophoric and dry air-stable; they can be stored for months without any change in reducing capacity. Stage II powders are easily handled in an open ambient environment, but readily react with water to produce stoichiometric yields of pure hydrogen gas. Stage II powders function well as a hydrogen source or drying agent.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service