Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655740

Sigma-Aldrich

Cyclohexylacetyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13ClO
CAS Number:
Molecular Weight:
160.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

bp

187 °F (lit.)

density

1.047 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

ClC(=O)CC1CCCCC1

InChI

1S/C8H13ClO/c9-8(10)6-7-4-2-1-3-5-7/h7H,1-6H2

InChI key

VABYVFZVTIDNOA-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

177.8 °F - closed cup

flash_point_c

81 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service